siRNA Details
Gene Symbol: Wls | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026582.4 | NP_080858.3 | 12 | 1796 | ||
GenBank mRNA | AK019886.1 | - | 13 | 1835 | ||
AK029053.1 | - | 12 | 1755 | |||
AK029104.1 | - | 12 | 1753 | |||
AK044810.1 | - | 12 | 1755 | |||
AK049842.1 | - | 12 | 1753 | |||
AK128912.1 | - | 12 | 1752 | |||
AK159207.1 | - | 12 | 1753 | |||
BC018381.1 | - | 11 | 1380 | 4237619 | ||
BC075615.1 | - | 12 | 1796 | 5703610 |
