siRNA Details
Gene Symbol: Rab14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026697.3 | NP_080973.1 | 4 | 613 | ||
GenBank mRNA | AB232609.1 | - | 3 | 244 | ||
AJ968955.1 | - | 4 | 468 | |||
AJ968956.1 | - | 3 | 414 | |||
AJ968957.1 | - | 4 | 468 | |||
AJ968958.1 | - | 3 | 414 | |||
AJ968959.1 | - | 4 | 468 | |||
AJ968960.1 | - | 3 | 414 | |||
AK002704.1 | - | 4 | 485 | |||
AK013410.1 | - | 4 | 466 | |||
AK083451.1 | - | 1 | 1484 | |||
AK133963.1 | - | 4 | 513 | |||
AK135751.1 | - | 4 | 502 | |||
AK160859.1 | - | 4 | 613 | |||
AK165812.1 | - | 4 | 460 | |||
AK168551.1 | - | 4 | 470 | |||
AK168867.1 | - | 4 | 482 | |||
AK169396.1 | - | 4 | 484 | |||
AK186545.1 | - | 1 | 21 | |||
AK216803.1 | - | 1 | 31 | |||
BC009085.1 | - | 4 | 296 | 2649852 | ||
BC025139.1 | - | 4 | 484 | 3980228 | ||
BC056648.1 | - | 4 | 486 | 5034519 |

More Information
Other Species Targeted

Gene Ontology Categories:
