siRNA Details
Gene Symbol: Vps51 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081041.1 | NP_001074510.1 | 10 | 2123 | ||
GenBank mRNA | AK003717.1 | - | 1 | 292 | ||
AK137155.1 | - | 10 | 2133 | |||
AK146211.1 | - | 10 | 2132 | |||
AK195187.1 | - | 3 | 140 | |||
AK212543.1 | - | 2 | 102 | |||
AK216884.1 | - | 3 | 140 |
