siRNA Details
Gene Symbol: Evc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145920.3 | NP_666032.1 | 4, 5 | 619 | ||
GenBank mRNA | AB083066.1 | - | 4, 5 | 549 | ||
AK044977.1 | - | 4, 5 | 490 | |||
AK134298.1 | - | 4, 5 | 602 | |||
AK165051.1 | - | 4, 5 | 504 | |||
BC037473.1 | - | 4, 5 | 600 | 4237472 | ||
BC064473.1 | - | 4, 5 | 610 | 3156380 |
