siRNA Details
Gene Symbol: Mthfd2l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026788.1 | NP_081064.1 | 8 | 1088 | ||
GenBank mRNA | AK003824.1 | - | 6 | 656 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Mthfd2l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026788.1 | NP_081064.1 | 8 | 1088 | ||
GenBank mRNA | AK003824.1 | - | 6 | 656 |