siRNA Details
Gene Symbol: Dcaf12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026893.3 | NP_081169.2 | 2 | 531 | ||
GenBank mRNA | AK005099.1 | - | 2 | 477 | ||
AK030173.1 | - | 2 | 483 | |||
AK031089.1 | - | 2 | 461 | |||
AK083369.1 | - | 1 | 1481 | |||
AK087517.1 | - | 2 | 414 | |||
AK090028.1 | - | 2 | 480 | |||
AK143635.1 | - | 2 | 484 | |||
AK173289.1 | - | 2 | 391 | |||
AK188223.1 | - | 1 | 39 | |||
BC023852.1 | - | 2 | 467 | 5346910 |
