siRNA Details
Gene Symbol: Med27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290489.1 | NP_001277418.1 | 7 | 661 | ||
NM_026896.5 | NP_081172.2 | 8 | 935 | |||
GenBank mRNA | AI850753.1 | - | 1 | 338 | ||
AK005251.1 | - | 8 | 910 | |||
AK088740.1 | - | 1 | 882 | |||
BC016537.1 | - | 8 | 926 | 3585027 |
