siRNA Details
Gene Symbol: Ascc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199187.2 | NP_001186116.1 | 10 | 1104 | ||
NM_026937.3 | NP_081213.1 | 10 | 1117 | |||
GenBank mRNA | AK007519.1 | - | 10 | 1101 | ||
AK139944.1 | - | 10 | 1104 | |||
AK171958.1 | - | 10 | 1078 | |||
AW457357.1 | - | 2 | 385 | |||
BC030905.1 | - | 10 | 1083 | 4503445 |
