siRNA Details
Gene Symbol: Lnp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110209.1 | NP_001103679.1 | 12 | 1123 | ||
NM_027133.3 | NP_081409.1 | 12 | 1123 | |||
GenBank mRNA | AK014897.1 | - | 12 | 1020 | ||
AK048501.1 | - | 3 | 1366 | |||
AK082900.1 | - | 11 | 1048 | |||
AK136506.1 | - | 12 | 1098 | |||
AK137445.1 | - | 12 | 1124 | |||
AK173241.1 | - | 12 | 1055 | |||
AY312281.1 | - | 12 | 1044 | |||
BC057961.1 | - | 12 | 1050 | 5353629 | ||
BC060153.1 | - | 11 | 962 |
