siRNA Details
Gene Symbol: Clybl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029556.3 | NP_083832.2 | 6 | 713 | ||
GenBank mRNA | AF428254.1 | - | 6 | 686 | ||
AK002741.1 | - | 6 | 841 | |||
AK009345.1 | - | 6 | 701 | |||
BC023398.1 | - | 6 | 703 | 5064709 |
