siRNA Details
Gene Symbol: Clybl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029556.3 | NP_083832.2 | 3 | 442 | ||
GenBank mRNA | AF428254.1 | - | 3 | 415 | ||
AK002741.1 | - | 3 | 568 | |||
AK009345.1 | - | 3 | 430 | |||
BC023398.1 | - | 3 | 432 | 5064709 |
