siRNA Details
Gene Symbol: Mllt3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286158.1 | NP_001273087.1 | 4, 5 | 419 | ||
NM_027326.3 | NP_081602.3 | 8, 9 | 1746 | |||
NM_029931.3 | NP_084207.2 | 4, 5 | 419 | |||
GenBank mRNA | AF333960.1 | - | 7, 8 | 1407 | ||
AK008707.1 | - | 4, 5 | 381 | |||
AK019458.1 | - | 4, 5 | 411 | |||
AK136066.1 | - | 4, 5 | 980 | |||
AK147009.1 | - | 4, 5 | 333 | |||
BC021420.1 | - | 9, 10 | 1585 | 5012671 | ||
BU614509.1 | - | 4, 5 | 547 | |||
BY713607.1 | - | 4, 5 | 407 | |||
DV051568.1 | - | 429 |
