siRNA Details
Gene Symbol: Mllt3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286158.1 | NP_001273087.1 | 3 | 253 | ||
NM_027326.3 | NP_081602.3 | 7 | 1580 | |||
NM_029931.3 | NP_084207.2 | 3 | 253 | |||
GenBank mRNA | AF333960.1 | - | 6 | 1241 | ||
AK008707.1 | - | 3 | 215 | |||
AK019458.1 | - | 3 | 245 | |||
AK136066.1 | - | 3 | 813 | |||
AK147009.1 | - | 3 | 167 | |||
BC021420.1 | - | 8 | 1419 | 5012671 | ||
BU614509.1 | - | 3 | 381 | |||
BY033742.1 | - | 3 | 248 | |||
BY713607.1 | - | 3 | 241 | |||
DV051568.1 | - | 263 |
