siRNA Details
Gene Symbol: Dpf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001267626.1 | NP_001254555.1 | 9 | 971 | ||
NM_058212.2 | NP_478119.1 | 9 | 968 | |||
GenBank mRNA | AF362750.1 | - | 9 | 940 | ||
AK012970.1 | - | 9 | 976 | |||
BC048572.1 | - | 9 | 851 | 6705809 | ||
FJ151218.1 | - | 9 | 943 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001191818, XM_008764748.
Gene Ontology Categories:
