siRNA Details
Gene Symbol: Gorasp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027352.4 | NP_081628.3 | 3, 4 | 537 | ||
GenBank mRNA | AK002515.1 | - | 4, 5 | 616 | ||
AK010664.2 | - | 3, 4 | 456 | |||
AK033413.1 | - | 3, 4 | 471 | |||
AK044691.1 | - | 422 | ||||
AK077153.1 | - | 3, 4 | 413 | |||
AK077683.1 | - | 3, 4 | 472 | |||
AK077706.1 | - | 3, 4 | 404 | |||
AK088239.1 | - | 3, 4 | 418 | |||
AK148337.1 | - | 3, 4 | 471 | |||
AK151843.1 | - | 3, 4 | 419 | |||
AK152475.1 | - | 3, 4 | 419 | |||
AK152544.1 | - | 3, 4 | 419 | |||
AK152603.1 | - | 3, 4 | 447 | |||
AK153049.1 | - | 3, 4 | 419 | |||
AK153555.1 | - | 3, 4 | 470 | |||
AK159469.1 | - | 3, 4 | 469 | |||
AK167823.1 | - | 3, 4 | 471 | |||
AK168928.1 | - | 4, 5 | 667 | |||
AK169382.1 | - | 3, 4 | 419 | |||
AK207507.1 | - | 1, 2 | 16 | |||
AK212581.1 | - | 1, 2 | 16 | |||
AK216052.1 | - | 1, 2 | 76 | |||
BC005600.1 | - | 3, 4 | 410 | 3482776 | ||
BC016455.1 | - | 2, 3 | 262 | 4234343 | ||
BC024982.1 | - | 3, 4 | 423 | |||
BU707592.1 | - | 3, 4 | 537 |
