siRNA Details
Gene Symbol: Gorasp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027352.4 | NP_081628.3 | 6 | 770 | ||
GenBank mRNA | AK002515.1 | - | 7 | 849 | ||
AK010664.2 | - | 6 | 689 | |||
AK033413.1 | - | 6 | 704 | |||
AK044691.1 | - | 655 | ||||
AK077153.1 | - | 6 | 645 | |||
AK077683.1 | - | 6 | 705 | |||
AK077706.1 | - | 6 | 637 | |||
AK088239.1 | - | 6 | 651 | |||
AK148337.1 | - | 6 | 704 | |||
AK151843.1 | - | 6 | 652 | |||
AK152475.1 | - | 6 | 652 | |||
AK152544.1 | - | 6 | 652 | |||
AK152603.1 | - | 6 | 680 | |||
AK153049.1 | - | 6 | 652 | |||
AK153555.1 | - | 6 | 703 | |||
AK159469.1 | - | 6 | 702 | |||
AK167823.1 | - | 6 | 704 | |||
AK168928.1 | - | 7 | 900 | |||
AK169382.1 | - | 6 | 652 | |||
AK184024.1 | - | 1 | 27 | |||
AK188100.1 | - | 1 | 27 | |||
AK197786.1 | - | 1 | 27 | |||
AK198884.1 | - | 1 | 27 | |||
AK216984.1 | - | 1 | 27 | |||
AK218436.1 | - | 1 | 27 | |||
BC005600.1 | - | 6 | 643 | 3482776 | ||
BC016455.1 | - | 5 | 495 | 4234343 | ||
BC024982.1 | - | 6 | 656 |
