siRNA Details
Gene Symbol: Hdac8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027382.3 | NP_081658.1 | 5 | 591 | ||
GenBank mRNA | AK011332.1 | - | 5 | 592 | ||
AK034511.1 | - | 5 | 557 | |||
AK041965.1 | - | 5 | 590 | |||
AK131998.1 | - | 5 | 804 | |||
BC061257.1 | - | 5 | 563 | 30251620 |
