siRNA Details
Gene Symbol: Gpatch2l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027405.2 | NP_081681.2 | 2, 3 | 733 | ||
GenBank mRNA | AK006185.1 | - | 2, 3 | 734 | ||
AK031712.1 | - | 2, 3 | 737 | |||
AK129299.1 | - | 2, 3 | 710 | |||
AK147916.1 | - | 2, 3 | 722 | |||
BC044824.1 | - | 1, 2 | 53 | |||
BC048169.1 | - | 2, 3 | 742 | 6821902 | ||
BC050782.1 | - | 2, 3 | 713 | |||
BC058256.1 | - | 2, 3 | 733 | 6466603 |
