siRNA Details
Gene Symbol: Tmtc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028651.2 | NP_082927.1 | 2 | 214 | ||
GenBank mRNA | AK028333.1 | - | 2 | 195 | ||
AK033947.1 | - | 2 | 215 | |||
AK036741.1 | - | 2 | 171 | |||
AK039093.1 | - | 2 | 159 | |||
AK043293.1 | - | 2 | 241 | |||
AK044391.1 | - | 2 | 170 | |||
AK045271.1 | - | 2 | 176 | |||
AK049748.1 | - | 3 | 355 | |||
AK082526.1 | - | 1 | 42 |
