siRNA Details
Gene Symbol: 5730455P16Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027472.3 | NP_081748.1 | 8 | 793 | ||
GenBank mRNA | AK017667.1 | - | 8 | 784 | ||
AK078548.1 | - | 8 | 794 | |||
AK135352.1 | - | 8 | 775 | |||
AK153246.1 | - | 8 | 792 | |||
AK160515.1 | - | 6 | 436 | |||
BC044897.1 | - | 8 | 781 | 3990240 | ||
BC055024.1 | - | 8 | 777 | 5149944 |
