siRNA Details
Gene Symbol: Ssfa2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080558.4 | NP_542125.3 | 12 | 2922 | ||
GenBank mRNA | AB093303.1 | - | 12 | 2797 | ||
AB120565.1 | - | 12 | 2807 | |||
AK037213.1 | - | 10 | 2632 | |||
AK169543.1 | - | 13 | 3100 | |||
BC008560.1 | - | 5 | 1895 | |||
BC018526.1 | - | 2 | 126 | |||
BC079535.1 | - | 6 | 2301 | |||
BC122519.1 | - | 12 | 2808 | |||
BC141883.1 | - | 12 | 2807 | 40097831 |
