siRNA Details
Gene Symbol: Dnajb14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033155.1 | NP_001028327.1 | 5 | 924 | ||
GenBank mRNA | AK139489.1 | - | 5 | 650 | ||
BC117717.1 | - | 5 | 725 | 40088109 | ||
BC117718.1 | - | 5 | 725 | 40088110 |
