siRNA Details
Gene Symbol: Sik3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027498.3 | NP_081774.3 | 19 | 2408 | ||
GenBank mRNA | AK129257.1 | - | 13 | 1421 | ||
BC058975.1 | - | 2 | 121 | |||
BC063268.1 | - | 19 | 2204 | 4187769 | ||
BC080688.1 | - | 19 | 2350 | 6515742 | ||
BC082313.1 | - | 5 | 563 |
