siRNA Details
Gene Symbol: Sik3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027498.3 | NP_081774.3 | 7 | 933 | ||
GenBank mRNA | AK017789.1 | - | 6 | 683 | ||
BC063268.1 | - | 7 | 729 | 4187769 | ||
BC080688.1 | - | 7 | 875 | 6515742 |
