siRNA Details
Gene Symbol: Glb1l3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113323.1 | NP_001106794.1 | 3 | 529 | ||
GenBank mRNA | AK014852.1 | - | 3 | 362 | ||
BC132200.1 | - | 3 | 322 | 40130477 | ||
BC132202.1 | - | 3 | 322 | 40130479 | ||
DV051729.1 | - | 3 | 549 |
