siRNA Details
Gene Symbol: Nol8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271397.1 | NP_001258326.1 | 2 | 683 | ||
GenBank mRNA | AK014994.1 | - | 2 | 684 | ||
AK029961.1 | - | 2 | 396 | |||
AK147172.1 | - | 2 | 419 | |||
BC049166.1 | - | 2 | 274 | |||
BC058699.1 | - | 2 | 839 | |||
BC125575.1 | - | 2 | 409 | 40130090 | ||
BC125579.1 | - | 2 | 409 | 40130094 |
