siRNA Details
Gene Symbol: Spef1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027641.2 | NP_081917.1 | 3 | 399 | ||
GenBank mRNA | AK016476.1 | - | 3 | 420 | ||
AK046964.1 | - | 465 | ||||
AK047675.1 | - | 3 | 400 | |||
AY860964.1 | - | 3 | 417 | |||
BC006046.1 | - | 3 | 339 | 3590641 |
