siRNA Details
Gene Symbol: Mgat4e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145300.2 | NP_001138772.1 | 3 | 1170 | ||
NM_001291015.1 | NP_001277944.1 | 4 | 1267 | |||
GenBank mRNA | AI561630.1 | - | 1 | 190 | ||
AK016512.1 | - | 3 | 1167 | |||
BC145149.1 | - | 3 | 824 | 9053700 | ||
BC145786.1 | - | 3 | 824 | 40131089 |
