siRNA Details
Gene Symbol: Asun | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138757.2 | NP_620096.2 | 16 | 2601 | ||
GenBank mRNA | AK016882.1 | - | 1 | 716 | ||
BC005696.1 | - | 4 | 562 | |||
BC026851.1 | - | 16 | 2530 | 4459471 | ||
BC027229.1 | - | 16 | 2186 | 2615769 | ||
BE685943.1 | - | 2 | 389 | |||
CF949591.1 | - | 4 | 544 |
