siRNA Details
Gene Symbol: Flrt3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172160.1 | NP_001165631.1 | 3 | 2376 | ||
NM_178382.4 | NP_848469.1 | 3 | 1823 | |||
GenBank mRNA | AK028252.1 | - | 3 | 1817 | ||
AK031464.1 | - | 3 | 1817 | |||
AK129369.1 | - | 3 | 2300 | |||
AK161100.1 | - | 3 | 1818 | |||
AY495670.1 | - | 1 | 1395 | |||
BC052043.1 | - | 3 | 1798 | 5702881 |
