siRNA Details
Gene Symbol: 9030624J02Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027815.4 | NP_082091.3 | 25 | 2574 | ||
GenBank mRNA | AK050302.1 | - | 25 | 2106 | ||
AK135468.1 | - | 1 | 47 | |||
BC007154.1 | - | 496 | ||||
BC025808.1 | - | 10 | 761 | |||
BC043674.1 | - | 18 | 1551 | 5044565 | ||
BC054720.1 | - | 25 | 2092 | 5689248 |
