siRNA Details
Gene Symbol: 9030624J02Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027815.4 | NP_082091.3 | 28, 29 | 3012 | ||
GenBank mRNA | AK050302.1 | - | 28, 29 | 2544 | ||
AK135468.1 | - | 4, 5 | 485 | |||
BC007154.1 | - | 934 | ||||
BC025808.1 | - | 13, 14 | 1199 | |||
BC038055.1 | - | 2, 3 | 279 | |||
BC043674.1 | - | 21, 22 | 1989 | 5044565 | ||
BC054720.1 | - | 28, 29 | 2530 | 5689248 |
