siRNA Details
Gene Symbol: 9030624J02Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027815.4 | NP_082091.3 | 3, 4 | 772 | ||
GenBank mRNA | AK011615.1 | - | 3, 4 | 282 | ||
AK018573.1 | - | 3, 4 | 312 | |||
AK050302.1 | - | 3, 4 | 304 | |||
AK136140.1 | - | 3, 4 | 312 | |||
BC054720.1 | - | 3, 4 | 290 | 5689248 |
