siRNA Details
Gene Symbol: Fuca1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024243.4 | NP_077205.3 | 1, 2 | 432 | ||
GenBank mRNA | AK002230.1 | - | 1, 2 | 400 | ||
AK053323.1 | - | 1, 2 | 377 | |||
AK089958.1 | - | 1, 2 | 402 | |||
AK150526.1 | - | 1, 2 | 367 | |||
AK150890.1 | - | 1, 2 | 424 | |||
AK151258.1 | - | 1, 2 | 400 | |||
AK151361.1 | - | 1, 2 | 421 | |||
AK151879.1 | - | 1, 2 | 421 | |||
AK151908.1 | - | 1, 2 | 375 | |||
AK152070.1 | - | 1, 2 | 375 | |||
AK165604.1 | - | 1, 2 | 402 | |||
AK170503.1 | - | 1, 2 | 412 | |||
BC003235.1 | - | 1, 2 | 405 | 3497631 |
