siRNA Details
Gene Symbol: Fuca1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024243.4 | NP_077205.3 | 5 | 977 | ||
GenBank mRNA | AK002230.1 | - | 5 | 945 | ||
AK053323.1 | - | 5 | 922 | |||
AK089958.1 | - | 5 | 947 | |||
AK150526.1 | - | 5 | 912 | |||
AK150890.1 | - | 5 | 967 | |||
AK151258.1 | - | 5 | 945 | |||
AK151361.1 | - | 5 | 966 | |||
AK151879.1 | - | 5 | 966 | |||
AK151908.1 | - | 5 | 920 | |||
AK152070.1 | - | 5 | 920 | |||
AK165604.1 | - | 5 | 947 | |||
AK170503.1 | - | 5 | 957 | |||
BC003235.1 | - | 5 | 950 | 3497631 |
