siRNA Details
Gene Symbol: Susd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001162913.1 | NP_001156385.1 | 7 | 1103 | ||
NM_027890.5 | NP_082166.3 | 9 | 1463 | |||
GenBank mRNA | AK004703.2 | - | 9 | 1433 | ||
AK028817.1 | - | 9 | 1439 | |||
AK033823.1 | - | 7 | 1076 | |||
AK037059.1 | - | 9 | 1445 | |||
AK037127.1 | - | 9 | 1414 | |||
AK038944.1 | - | 7 | 1731 | |||
AK042469.1 | - | 9 | 1447 | |||
AK128959.1 | - | 9 | 1435 | |||
AK167155.1 | - | 8 | 1645 | |||
BC026837.1 | - | 9 | 1416 | 4222578 | ||
BC066833.1 | - | 6 | 1262 |
