siRNA Details
Gene Symbol: Cul2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029402.3 | NP_083678.2 | 12, 13 | 1359 | ||
GenBank mRNA | AK004875.2 | - | 12, 13 | 1315 | ||
AK016520.1 | - | 12, 13 | 1329 | |||
AK028589.1 | - | 12, 13 | 1328 | |||
AK144693.1 | - | 12, 13 | 1333 | |||
AK160597.1 | - | 12, 13 | 1344 | |||
AK220287.1 | - | 12, 13 | 1334 | |||
BC025902.1 | - | 4, 5 | 425 | |||
BC026779.1 | - | 12, 13 | 1260 | 4018298 | ||
BC027428.1 | - | 12, 13 | 1322 | 3983461 |
