siRNA Details
Gene Symbol: Cul2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029402.3 | NP_083678.2 | 10, 11 | 1185 | ||
GenBank mRNA | AK004875.2 | - | 10, 11 | 1141 | ||
AK016520.1 | - | 10, 11 | 1155 | |||
AK028589.1 | - | 10, 11 | 1154 | |||
AK144693.1 | - | 10, 11 | 1159 | |||
AK160597.1 | - | 10, 11 | 1170 | |||
AK220287.1 | - | 10, 11 | 1160 | |||
BC025902.1 | - | 2, 3 | 251 | |||
BC026779.1 | - | 10, 11 | 1086 | 4018298 |
