siRNA Details
Gene Symbol: Klhdc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163729.2 | NP_001157201.2 | 6 | 930 | ||
NM_027910.3 | NP_082186.2 | 7 | 1083 | |||
GenBank mRNA | AB053465.1 | - | 7 | 954 | ||
AJ278127.1 | - | 7 | 950 | |||
AK004964.1 | - | 7 | 1060 | |||
AK079082.1 | - | 7 | 961 | |||
AK141673.1 | - | 6 | 915 | |||
AK170623.1 | - | 7 | 955 | |||
AK186202.1 | - | 1 | 112 | |||
AK215007.1 | - | 1 | 42 | |||
BC018154.1 | - | 7 | 1052 | 3983436 |
