siRNA Details
Gene Symbol: Esco2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028039.2 | NP_082315.3 | 9 | 1632 | ||
GenBank mRNA | AA881675.1 | - | 1 | 114 | ||
AK144685.1 | - | 9 | 1588 | |||
BC033303.1 | - | 9 | 1547 | 3675568 | ||
BC071275.1 | - | 9 | 1565 | 6331551 |
