siRNA Details
Gene Symbol: 2610008E11Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004362.2 | NP_001004362.2 | 6 | 778 | ||
GenBank mRNA | AK148309.1 | - | 6 | 779 | ||
BC082302.1 | - | 6 | 735 | 6838422 |
