siRNA Details
Gene Symbol: Fbxl20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028149.1 | NP_082425.1 | 10 | 1000 | ||
GenBank mRNA | AK012109.1 | - | 10 | 724 | ||
AK144786.1 | - | 10 | 974 | |||
AK220232.1 | - | 10 | 953 | |||
EF649694.1 | - | 10 | 747 |
