siRNA Details
Gene Symbol: Palld | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081390.1 | NP_001074859.1 | 9 | 1792 | ||
NM_001293772.1 | NP_001280701.1 | 9 | 1796 | |||
NM_001293773.1 | NP_001280702.1 | 8 | 927 | |||
GenBank mRNA | AF205079.1 | - | 3 | 220 | ||
AK031696.1 | - | 8 | 919 | |||
AK052489.1 | - | 9 | 1774 | |||
AK076423.1 | - | 7 | 564 | |||
AK087270.1 | - | 8 | 929 | |||
AK173081.1 | - | 8 | 867 | |||
BC027364.1 | - | 8 | 876 | |||
JX477684.1 | - | 8 | 919 |
