siRNA Details
Gene Symbol: Palld | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081390.1 | NP_001074859.1 | 5, 6 | 1512 | ||
NM_001293772.1 | NP_001280701.1 | 5, 6 | 1516 | |||
NM_001293773.1 | NP_001280702.1 | 4, 5 | 647 | |||
GenBank mRNA | AK031696.1 | - | 4, 5 | 639 | ||
AK052489.1 | - | 5, 6 | 1494 | |||
AK076423.1 | - | 3, 4 | 284 | |||
AK087270.1 | - | 4, 5 | 649 | |||
AK162050.1 | - | 4, 5 | 639 | |||
AK173081.1 | - | 4, 5 | 587 | |||
BC027364.1 | - | 4, 5 | 596 | |||
JX477684.1 | - | 4, 5 | 639 |
