siRNA Details
Gene Symbol: Hnrnpll | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144802.4 | NP_659051.3 | 5 | 1086 | ||
GenBank mRNA | AK012866.1 | - | 5 | 802 | ||
AK034860.1 | - | 5 | 1031 | |||
AK048900.1 | - | 5 | 875 | |||
AK146879.1 | - | 5 | 1048 | |||
AK160943.1 | - | 4 | 894 | |||
AK171091.1 | - | 5 | 953 | |||
BC004763.1 | - | 2 | 59 | |||
BC012849.1 | - | 5 | 1032 | 3587716 |
