siRNA Details
Gene Symbol: Prdm4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK028414.1 | - | 8, 9 | 1805 | ||
AK029718.1 | - | 5, 6 | 1262 | |||
BC025642.1 | - | 5, 6 | 1317 | |||
BC042516.1 | - | 6, 7 | 1502 | |||
BC057596.1 | - | 8, 9 | 1695 | |||
BC139409.1 | - | 7, 8 | 1685 | 8862431 | ||
BC145619.1 | - | 7, 8 | 1664 | 9054187 |
