siRNA Details
Gene Symbol: Prdm4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK013393.1 | - | 3 | 693 | ||
AK028414.1 | - | 12 | 2672 | |||
AK029718.1 | - | 9 | 2129 | |||
BC025642.1 | - | 9 | 2184 | |||
BC042516.1 | - | 10 | 2369 | |||
BC057596.1 | - | 12 | 2562 | |||
BC139409.1 | - | 11 | 2552 | 8862431 | ||
BC145619.1 | - | 11 | 2531 | 9054187 |
