siRNA Details
Gene Symbol: Prdm4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK028414.1 | - | 8 | 1743 | ||
AK029718.1 | - | 5 | 1200 | |||
BC025642.1 | - | 5 | 1255 | |||
BC042516.1 | - | 6 | 1440 | |||
BC057596.1 | - | 8 | 1633 | |||
BC139409.1 | - | 7 | 1623 | 8862431 | ||
BC145619.1 | - | 7 | 1602 | 9054187 |
