siRNA Details
Gene Symbol: Prrc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028447.3 | NP_082723.2 | 6 | 1079 | ||
GenBank mRNA | AK004443.1 | - | 6 | 1079 | ||
AK143537.1 | - | 6 | 1071 | |||
BC014817.1 | - | 6 | 995 | 3709357 |
