siRNA Details
Gene Symbol: Rassf6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028478.2 | NP_082754.2 | 9, 10 | 990 | ||
GenBank mRNA | AK005472.1 | - | 9, 10 | 938 | ||
AK132288.1 | - | 9, 10 | 952 | |||
BC031808.1 | - | 9, 10 | 962 | |||
BC039934.1 | - | 9, 10 | 1037 | |||
BC075725.1 | - | 6, 7 | 2328 |
